Anti-Complexin-1/2 Mouse Monoclonal Antibody [clone: 837802]

Supplier: R & D Systems
MAB8069
102887-454EA 577.02 USD
102887-454
Anti-Complexin-1/2 Mouse Monoclonal Antibody [clone: 837802]
Antibodies
Simple Western

Type: Primary
Antigen Symbol:
Antigen Name: Complexin-1/2
Antigen Synonyms:
Clonality: Monoclonal
Clone: 837802
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR