Anti-DAB2 Mouse Monoclonal Antibody [clone: 883216]

Supplier: R & D Systems
MAB8064
102887-450EA 577.02 USD
102887-450
Anti-DAB2 Mouse Monoclonal Antibody [clone: 883216]
Antibodies
Type: Primary
Antigen: DAB2
Clonality: Monoclonal
Clone: 883216
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR