Anti-EGFR Rat Monoclonal Antibody [clone: 869201]

Supplier: R & D Systems
MAB8058
102887-448EA 577.02 USD
102887-448
Anti-EGFR Rat Monoclonal Antibody [clone: 869201]
Antibodies
Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: 869201
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR