Anti-STK11 Mouse Monoclonal Antibody [clone: 873502]

Supplier: R & D Systems
MAB8055
102887-444EA 577.02 USD
102887-444
Anti-STK11 Mouse Monoclonal Antibody [clone: 873502]
Antibodies
Type: Primary
Antigen: STK11
Clonality: Monoclonal
Clone: 873502
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR