Anti-BATF Mouse Monoclonal Antibody [clone: 687706]

Supplier: R & D Systems
MAB8054
102887-442EA 577.02 USD
102887-442
Anti-BATF Mouse Monoclonal Antibody [clone: 687706]
Antibodies
Type: Primary
Antigen: BATF
Clonality: Monoclonal
Clone: 687706
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR