Anti-IL1B Mouse Monoclonal Antibody [clone: 855403]

Supplier: R & D Systems
MAB8049
102887-440EA 763.57 USD
102887-440
Anti-IL1B Mouse Monoclonal Antibody [clone: 855403]
Antibodies
Type: Primary
Antigen: IL1B
Clonality: Monoclonal
Clone: 855403
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Horse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR