Anti-EEA1 Mouse Monoclonal Antibody [clone: 871546]

Supplier: R & D Systems
MAB8047
102887-436EA 594.37 USD
102887-436
Anti-EEA1 Mouse Monoclonal Antibody [clone: 871546]
Antibodies
Type: Primary
Antigen: EEA1
Clonality: Monoclonal
Clone: 871546
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR