Anti-OTOL1 Mouse Monoclonal Antibody [clone: 839418]

Supplier: R & D Systems
MAB8045
102887-434EA 577.02 USD
102887-434
Anti-OTOL1 Mouse Monoclonal Antibody [clone: 839418]
Antibodies
Type: Primary
Antigen: OTOL1
Clonality: Monoclonal
Clone: 839418
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR