Anti-IGFBP4 Mouse Monoclonal Antibody [clone: 82314]

Supplier: R & D Systems
MAB8041
220021-566EA 779.48 USD
220021-566
Anti-IGFBP4 Mouse Monoclonal Antibody [clone: 82314]
Antibodies
Type: Primary
Antigen: IGFBP4
Clonality: Monoclonal
Clone: 82314
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR