Anti-HOXB7 Mouse Monoclonal Antibody [clone: 864617]

Supplier: R & D Systems
MAB80401
102887-432EA 561.11 USD
102887-432
Anti-HOXB7 Mouse Monoclonal Antibody [clone: 864617]
Antibodies
Type: Primary
Antigen: HOXB7
Clonality: Monoclonal
Clone: 864617
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR