Anti-IGFBP4 Mouse Monoclonal Antibody [clone: 82334]

Supplier: R & D Systems
MAB804
220021-565EA 763.57 USD
220021-565
Anti-IGFBP4 Mouse Monoclonal Antibody [clone: 82334]
Antibodies
Type: Primary
Antigen: IGFBP4
Clonality: Monoclonal
Clone: 82334
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR