Anti-RNF31 Mouse Monoclonal Antibody [clone: 875227]

Supplier: R & D Systems
MAB8039
102887-428EA 639.2 USD
102887-428
Anti-RNF31 Mouse Monoclonal Antibody [clone: 875227]
Antibodies
Simple Western

Type: Primary
Antigen: RNF31
Clonality: Monoclonal
Clone: 875227
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR