Anti-SART1 Mouse Monoclonal Antibody [clone: 865709]

Supplier: R & D Systems
MAB8034
102887-426EA 577.02 USD
102887-426
Anti-SART1 Mouse Monoclonal Antibody [clone: 865709]
Antibodies
Type: Primary
Antigen: SART1
Clonality: Monoclonal
Clone: 865709
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR