Anti-PARD3 Mouse Monoclonal Antibody [clone: 833517]

Supplier: R & D Systems
MAB8030
102887-424EA 594.37 USD
102887-424
Anti-PARD3 Mouse Monoclonal Antibody [clone: 833517]
Antibodies
Type: Primary
Antigen: PARD3
Clonality: Monoclonal
Clone: 833517
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR