Anti-SQSTM1 Mouse Monoclonal Antibody [clone: 864807]

Supplier: R & D Systems
MAB8028
102887-422EA 655.11 USD
102887-422
Anti-SQSTM1 Mouse Monoclonal Antibody [clone: 864807]
Antibodies
Simple Western

Type: Primary
Antigen: SQSTM1
Clonality: Monoclonal
Clone: 864807
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR