Anti-GLUD1 / GLUD2 Mouse Monoclonal Antibody [clone: 848209]

Supplier: R & D Systems
MAB8027
102887-420EA 594.37 USD
102887-420
Anti-GLUD1 / GLUD2 Mouse Monoclonal Antibody [clone: 848209]
Antibodies
Simple Western

Type: Primary
Antigen Symbol: GLUD1/2
Antigen Name:
Antigen Synonyms:
Clonality: Monoclonal
Clone: 848209
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR