Anti-MLANA Mouse Monoclonal Antibody [clone: 872719]

Supplier: R & D Systems
MAB8008
102887-416EA 594.37 USD
102887-416
Anti-MLANA Mouse Monoclonal Antibody [clone: 872719]
Antibodies
Type: Primary
Antigen: MLANA
Clonality: Monoclonal
Clone: 872719
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR