Anti-ITLN2 Mouse Monoclonal Antibody [clone: 874029]

Supplier: R & D Systems
MAB8004
102887-414EA 584.25 USD
102887-414
Anti-ITLN2 Mouse Monoclonal Antibody [clone: 874029]
Antibodies
Type: Primary
Antigen: ITLN2
Clonality: Monoclonal
Clone: 874029
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR