Anti-TM4SF4 Mouse Monoclonal Antibody [clone: 832441]

Supplier: R & D Systems
MAB7998
102887-412EA 594.37 USD
102887-412
Anti-TM4SF4 Mouse Monoclonal Antibody [clone: 832441]
Antibodies
Type: Primary
Antigen: TM4SF4
Clonality: Monoclonal
Clone: 832441
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR