Anti-MIP-I Mouse Monoclonal Antibody [clone: 84420]

Supplier: R & D Systems
MAB799
220021-563EA 763.57 USD
220021-563
Anti-MIP-I Mouse Monoclonal Antibody [clone: 84420]
Antibodies
Type: Primary
Antigen: MIP-I
Clonality: Monoclonal
Clone: 84420
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Viral
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR