Anti-SIGLEC8 Mouse Monoclonal Antibody [clone: 837535]

Supplier: R & D Systems
MAB7975
102887-410EA 600.15 USD
102887-410
Anti-SIGLEC8 Mouse Monoclonal Antibody [clone: 837535]
Antibodies
Type: Primary
Antigen: SIGLEC8
Clonality: Monoclonal
Clone: 837535
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR