Anti-IGFBP2 Rat Monoclonal Antibody [clone: 150102]

Supplier: R & D Systems
MAB7971
220021-562EA 763.57 USD
220021-562
Anti-IGFBP2 Rat Monoclonal Antibody [clone: 150102]
Antibodies
Type: Primary
Antigen: IGFBP2
Clonality: Monoclonal
Clone: 150102
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR