Anti-IGFBP2 Rat Monoclonal Antibody [clone: 150101]

Supplier: R & D Systems
MAB797
220021-561EA 763.57 USD
220021-561
Anti-IGFBP2 Rat Monoclonal Antibody [clone: 150101]
Antibodies
Type: Primary
Antigen: IGFBP2
Clonality: Monoclonal
Clone: 150101
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR