Anti-RPS6KA1 (phospho-Ser380) Rabbit Monoclonal Antibody [clone: 1024A]

Supplier: R & D Systems
MAB79671
102887-408EA 614.62 USD
102887-408
Anti-RPS6KA1 (phospho-Ser380) Rabbit Monoclonal Antibody [clone: 1024A]
Antibodies
Simple Western

Type: Primary
Antigen Symbol: RPS6KA1
Antigen Name: ribosomal protein S6 kinase, polypeptide A1
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 1024A
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR