Anti-CSNK2A1 Mouse Monoclonal Antibody [clone: 844720]

Supplier: R & D Systems
MAB7957
102887-404EA 594.37 USD
102887-404
Anti-CSNK2A1 Mouse Monoclonal Antibody [clone: 844720]
Antibodies
Simple Western

Type: Primary
Antigen: CSNK2A1
Clonality: Monoclonal
Clone: 844720
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR