Anti-PF4 Mouse Monoclonal Antibody [clone: 170138]

Supplier: R & D Systems
MAB7952
220021-559EA 623.29 USD
220021-559
Anti-PF4 Mouse Monoclonal Antibody [clone: 170138]
Antibodies
Type: Primary
Antigen: PF4
Clonality: Monoclonal
Clone: 170138
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR