Anti-PF4 Mouse Monoclonal Antibody [clone: 170106]

Supplier: R & D Systems
MAB795
220021-557EA 772.25 USD
220021-557
Anti-PF4 Mouse Monoclonal Antibody [clone: 170106]
Antibodies
Simple Western

Type: Primary
Antigen: PF4
Clonality: Monoclonal
Clone: 170106
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR