Anti-TGOLN1 Rat Monoclonal Antibody [clone: 831629]

Supplier: R & D Systems
MAB7944
102887-400EA 594.37 USD
102887-400
Anti-TGOLN1 Rat Monoclonal Antibody [clone: 831629]
Antibodies
Type: Primary
Antigen: TGOLN1
Clonality: Monoclonal
Clone: 831629
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR