Anti-MCHR1 Mouse Monoclonal Antibody [clone: 851047]

Supplier: R & D Systems
MAB7938
102887-164EA 594.37 USD
102887-164
Anti-MCHR1 Mouse Monoclonal Antibody [clone: 851047]
Antibodies
Simple Western

Type: Primary
Antigen: MCHR1
Clonality: Monoclonal
Clone: 851047
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR