Anti-SLC30A8 Mouse Monoclonal Antibody [clone: 815039]

Supplier: R & D Systems
MAB7936
102887-162EA 594.37 USD
102887-162
Anti-SLC30A8 Mouse Monoclonal Antibody [clone: 815039]
Antibodies
Type: Primary
Antigen: SLC30A8
Clonality: Monoclonal
Clone: 815039
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR