Anti-PCDH12 Rat Monoclonal Antibody [clone: 868909]

Supplier: R & D Systems
MAB7926
102887-158EA 577.02 USD
102887-158
Anti-PCDH12 Rat Monoclonal Antibody [clone: 868909]
Antibodies
Type: Primary
Antigen: PCDH12
Clonality: Monoclonal
Clone: 868909
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR