Anti-GCHFR Mouse Monoclonal Antibody [clone: 846011]

Supplier: R & D Systems
MAB7918
102887-156EA 561.11 USD
102887-156
Anti-GCHFR Mouse Monoclonal Antibody [clone: 846011]
Antibodies
Simple Western

Type: Primary
Antigen Symbol: GCHFR
Antigen Name: GTP cyclohydrolase I feedback regulator
Antigen Synonyms: GFRP, HsT16933, P35
Clonality: Monoclonal
Clone: 846011
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR