Anti-MSX2 Mouse Monoclonal Antibody [clone: 786607]

Supplier: R & D Systems
MAB7917
102887-154EA 594.37 USD
102887-154
Anti-MSX2 Mouse Monoclonal Antibody [clone: 786607]
Antibodies
Type: Primary
Antigen: MSX2
Clonality: Monoclonal
Clone: 786607
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR