Anti-PTPRN Mouse Monoclonal Antibody [clone: 815811]

Supplier: R & D Systems
MAB7906
102887-152EA 594.37 USD
102887-152
Anti-PTPRN Mouse Monoclonal Antibody [clone: 815811]
Antibodies
Simple Western

Type: Primary
Antigen: PTPRN
Clonality: Monoclonal
Clone: 815811
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR