Anti-FCRLA Rat Monoclonal Antibody [clone: 865910]

Supplier: R & D Systems
MAB7905
102887-150EA 561.11 USD
102887-150
Anti-FCRLA Rat Monoclonal Antibody [clone: 865910]
Antibodies
Type: Primary
Antigen Symbol: FCRLA
Antigen Name: Fc receptor-like A
Antigen Synonyms: BB219290, FCRL, FCRL1, FREB, Fcrlm1, Fcrx, Freb1, mFREB, mFcrX
Clonality: Monoclonal
Clone: 865910
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR