Anti-DOC2A Mouse Monoclonal Antibody [clone: 842213]

Supplier: R & D Systems
MAB7904
102887-148EA 561.11 USD
102887-148
Anti-DOC2A Mouse Monoclonal Antibody [clone: 842213]
Antibodies
Type: Primary
Antigen: DOC2A
Clonality: Monoclonal
Clone: 842213
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR