Anti-CNN1 Mouse Monoclonal Antibody [clone: 836701]

Supplier: R & D Systems
MAB7900
102887-146EA 577.02 USD
102887-146
Anti-CNN1 Mouse Monoclonal Antibody [clone: 836701]
Antibodies
Type: Primary
Antigen: CNN1
Clonality: Monoclonal
Clone: 836701
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR