Anti-TCN2 Mouse Monoclonal Antibody [clone: 874420]

Supplier: R & D Systems
MAB7895
102887-142EA 577.02 USD
102887-142
Anti-TCN2 Mouse Monoclonal Antibody [clone: 874420]
Antibodies
Simple Western

Type: Primary
Antigen: TCN2
Clonality: Monoclonal
Clone: 874420
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR