Anti-STX4 Mouse Monoclonal Antibody [clone: 884637]

Supplier: R & D Systems
MAB7894
102887-140EA 577.02 USD
102887-140
Anti-STX4 Mouse Monoclonal Antibody [clone: 884637]
Antibodies
Simple Western

Type: Primary
Antigen: STX4
Clonality: Monoclonal
Clone: 884637
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR