Anti-TAGLN Mouse Monoclonal Antibody [clone: 859112]

Supplier: R & D Systems
MAB78861
102887-138EA 594.37 USD
102887-138
Anti-TAGLN Mouse Monoclonal Antibody [clone: 859112]
Antibodies
Simple Western

Type: Primary
Antigen: TAGLN
Clonality: Monoclonal
Clone: 859112
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR