Anti-TAGLN Mouse Monoclonal Antibody [clone: 859108]

Supplier: R & D Systems
MAB7886
102887-136EA 577.02 USD
102887-136
Anti-TAGLN Mouse Monoclonal Antibody [clone: 859108]
Antibodies
Type: Primary
Antigen: TAGLN
Clonality: Monoclonal
Clone: 859108
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR