Anti-MSTN Rat Monoclonal Antibody [clone: 84231]

Supplier: R & D Systems
MAB7881
220021-553EA 577.02 USD
220021-553
Anti-MSTN Rat Monoclonal Antibody [clone: 84231]
Antibodies
Type: Primary
Antigen: MSTN
Clonality: Monoclonal
Clone: 84231
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR