Anti-ZC3H12A Mouse Monoclonal Antibody [clone: 604421]

Supplier: R & D Systems
MAB7875
102887-130EA 647.88 USD
102887-130
Anti-ZC3H12A Mouse Monoclonal Antibody [clone: 604421]
Antibodies
Type: Primary
Antigen: ZC3H12A
Clonality: Monoclonal
Clone: 604421
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR