Anti-HIF1AN Mouse Monoclonal Antibody [clone: 745122]

Supplier: R & D Systems
MAB7874
102887-128EA 577.02 USD
102887-128
Anti-HIF1AN Mouse Monoclonal Antibody [clone: 745122]
Antibodies
Simple Western

Type: Primary
Antigen: HIF1AN
Clonality: Monoclonal
Clone: 745122
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR