Anti-THBS4 Rat Monoclonal Antibody [clone: 893655]

Supplier: R & D Systems
MAB7860
102887-398EA 594.37 USD
102887-398
Anti-THBS4 Rat Monoclonal Antibody [clone: 893655]
Antibodies
Type: Primary
Antigen: THBS4
Clonality: Monoclonal
Clone: 893655
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR