Anti-ARHGEF9 Mouse Monoclonal Antibody [clone: 809309]

Supplier: R & D Systems
MAB7848
102887-396EA 577.02 USD
102887-396
Anti-ARHGEF9 Mouse Monoclonal Antibody [clone: 809309]
Antibodies
Type: Primary
Antigen: ARHGEF9
Clonality: Monoclonal
Clone: 809309
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR