Anti-IL2RG Rat Monoclonal Antibody [clone: 644337]

Supplier: R & D Systems
MAB7843
102887-392EA 577.02 USD
102887-392
Anti-IL2RG Rat Monoclonal Antibody [clone: 644337]
Antibodies
Type: Primary
Antigen: IL2RG
Clonality: Monoclonal
Clone: 644337
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR