Anti-IL2RG Rat Monoclonal Antibody [clone: 644346]

Supplier: R & D Systems
MAB7842
102887-390EA 577.02 USD
102887-390
Anti-IL2RG Rat Monoclonal Antibody [clone: 644346]
Antibodies
Type: Primary
Antigen: IL2RG
Clonality: Monoclonal
Clone: 644346
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR