Anti-IL2RG Rat Monoclonal Antibody [clone: 135727]

Supplier: R & D Systems
MAB7841
220021-550EA 772.25 USD
220021-550
Anti-IL2RG Rat Monoclonal Antibody [clone: 135727]
Antibodies
Type: Primary
Antigen: IL2RG
Clonality: Monoclonal
Clone: 135727
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR