Anti-GCK Mouse Monoclonal Antibody [clone: 849520]

Supplier: R & D Systems
MAB7840
102887-388EA 594.37 USD
102887-388
Anti-GCK Mouse Monoclonal Antibody [clone: 849520]
Antibodies
Type: Primary
Antigen Symbol: GCK
Antigen Name: glucokinase (hexokinase 4)
Antigen Synonyms: FGQTL3, GK, GLK, HHF3, HK4, HKIV, HXKP, LGLK, MODY2
Clonality: Monoclonal
Clone: 849520
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR